Corning® Polystyrene Tissue Culture Flasks with Vented Caps, Sterile, Corning
Supplier: Corning
Certificates
About this item
These flasks are tissue culture-treated for optimum cell attachment and growth
- Certified nonpyrogenic
- Lot number is printed on each flask for traceability
- Canted flasks have a skirt to prevent tipping
The triangular flasks have the shape for increased stability and pipet access. The flasks packaged in cases of 84 come prelabeled with a bar code and is validated for use with Select Robotic System.
Polyethylene caps contain a 0.2 µm hydrophobic filter to allow gas exchange and minimize risk of contamination.
Specifications
- Catalog No:
- 89090-950
- 29186-010
- 29560-959
- 10010-366
- 29186-106
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta