


Cobalt(II) chloride hexahydrate 98.0-102.0% (by EDTA titration), crystals, AR® ACS, Macron Fine Chemicals™
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:7791-13-1
:
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652
:237.93 g/mol
Merck Index:13,02462
Boiling Pt:110 °C (1013 hPa)
Melting Pt:56 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3288
ADR:9,III
:Management of Change (MOC) category = N
EINECS:231-589-4
:GHS06
:GHS08
:GHS09
Specification Test Results
- ACS - Assay (CoCl₂·6H₂O) (by EDTA titrn):98.0 - 102.0 %
- ACS - Appearance (rose-red crystals):Passes Test
- ACS - Copper (Cu):≤0.002 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Iron (Fe):≤0.005 %
- ACS - Nickel (Ni):≤0.1 %
- ACS - Nitrate (NO₃):≤0.01 %
- ACS - Sulfate (SO₄):≤0.01 %
- ACS - Zinc (Zn):≤0.03 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.05 %
- ACS - Calcium (Ca):≤0.005 %
Specification Test Results
Related Information
Certificates
Search by Lot Number4532 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals