AlphaTec® 87-208 Orange Heavyweight Natural Rubber Latex Gloves, Ansell
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificates
About this item
These extra-heavy-duty gloves feature increased puncture/cut resistance and tensile strength to provide added protection against chemical and physical hazards.
- Hand-specific
- Straight cuffs
- Diamond-embossed finish
Cotton flock lining increases user comfort. For chemical processing, petroleum refineries, plant maintenance, and heavy equipment assembly and cleanup.
Caution: This product contains natural rubber latex, which may cause allergic reactions in some individuals.
Specifications
- Catalog No:
- 47751-224
- 47751-222
- 47747-400
- 47747-402
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Latex/Latex-free:
- Latex
- Latex