


Chloramine-T (sodium salt) trihydrate, J.T.Baker®
Catalog # JTE494-6
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Size:250 g
- Cat. No.:JTE494-6
- Supplier no.:E494-06
Specifications
Specification Test Results
- Active Chlorine:≥11 %
Specification Test Results
About this item
:
Merck Index:13,02085
Melting Pt:167…170 °C
Density:1.43 g/cm³ (20 °C)
EINECS:204-854-7
ADR:8,III
:Management of Change (MOC) category = N
Flash Pt:192 °C (closed cup)
:281.69 g/mol
UN:3263
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:7080-50-4
Formula:CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number:MFCD00149066