
Citric acid 99.5-100.5%, powder USP, Multi-Compendial, GMP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Regulated
About this item
CAS Number:77-92-9
:
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669
:192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.67 g/cm³
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5 %
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification:Passes Test
- USP - Residue on Ignition:≤0.1 %
- USP - Oxalic Acid:≤0.036 %
- USP - Sulfate (SO₄):≤0.015 %
- USP - Readily Carbonizable Substances:Passes Test
- USP - Identification C: Water (H₂O):≤1.0 %
- EP/BP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5 %
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Readily Carbonizable Substances:Passes Test
- EP/BP - Oxalic Acid:≤360 ppm
- EP/BP - Sulfate (SO₄):≤150 ppm
- EP/BP - Water (H₂O):≤1.0 %
- EP/BP - Ash (sulfated):≤0.1 %
- EP/BP - Endotoxin Concentration, <0.5 IU/mg:Passes Test
- JP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Identification:Passes Test
- JP - Sulfate (SO₄):≤150 ppm
- JP - Oxalic Acid:≤360 ppm
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Readily Carbonizable Substances:Passes Test
- JP - Residue on Ignition:≤0.10 %
- JP - Water (H₂O):≤1.0 %
- E. coli:Negative
- P. aureginosa:Negative
Specification Test Results
Related Information
Certificates
Search by Lot Number0127 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT0127-5 spec_JT0127-7