
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), crystalline powder FCC, USP, J.T.Baker®
Catalog # JT0120-5
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
MOC: RegulatedCertificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Size:2.5 kg
- Cat. No.:JT0120-5
- Supplier no.:0120-05
Specifications
Specification Test Results
- USP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5 %
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification (USP):Passes Test
- USP - Water (H₂O) (U.S.P.):7.5 - 9.0 %
- USP - Residue on Ignition:≤0.05 %
- USP - Oxalic Acid (USP):≤0.036 %
- USP - Sulfate (SO₄):≤0.015 %
- USP - Readily Carbonizable Substances:Passes Test
- FCC - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5 %
- FCC - Identification (FCC):Passes Test
- FCC - Lead (Pb):≤0.5 mg/kg
- FCC - Oxalate (C₂O₄) (FCC):≤100 mg/kg
- FCC - Readily Carbonizable Substances:Passes Test
- FCC - Residue on Ignition:≤0.05 %
- FCC - Water (H₂O) (F.C.C.):≤8.8 %
Specification Test Results
About this item
:
:210.14 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:135…152 °C
Melting Pt:135 - 152 °C
Density:1.552 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Flash Pt:173.9 °C (closed cup)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
:GHS07
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972