Anti-PI 3 Kinase p85 beta Rabbit Polyclonal Antibody
Catalog # 77437-638
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:PI 3 Kinase p85 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:18709
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Storage Temperature:Store at –20 °C for 12 months.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77437-638
- Packaging:Vial
Specifications
About this item
Binds to activated (phosphorylated) protein-tyrosine kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane.
Type: Primary
Antigen: PI 3 Kinase p85 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat, Bovine, Dog