Anti-SLC23A3 Rabbit Polyclonal Antibody
Catalog # 77439-330
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:SLC23A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:151295
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:65
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77439-330
- Packaging:Vial
Specifications
About this item
SLC23A3 (Solute Carrier Family 23, Member 3) is a Protein Coding gene. Diseases associated with SLC23A3 include epilepsy, generalized, with febrile seizures plus, type 5. GO annotations related to this gene include transporter activity. An important paralog of this gene is SLC23A1.
Type: Primary
Antigen: SLC23A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse