Anti-ZNF596 Rabbit Polyclonal Antibody
Catalog # 77438-722
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:ZNF596
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Gene ID:169270
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:55
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77438-722
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ZNF596
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human