Anti-Neural retinal specific leucine zipper/NRL Rabbit Polyclonal Antibody
Catalog # 77438-666
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Neural retinal specific leucine zipper/NRL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:4901
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Storage Temperature:Store at –20 °C for 12 months.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77438-666
- Packaging:Vial
Specifications
About this item
Transcription factor which regulates the expression of several rod-specific genes, in cluding RHO and PDE6B.
Type: Primary
Antigen: Neural retinal specific leucine zipper/NRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat