Order Entry
Puerto Rico
ContactUsLinkComponent
Chloroform ≥99.8%, OmniPur® for biotechnology
Toxic
Health
Chloroform ≥99.8%, OmniPur® for biotechnology
Catalog # EM-3150
Supplier:  MilliporeSigma
CAS Number:  
undefined
Toxic
Health
Chloroform ≥99.8%, OmniPur® for biotechnology
Catalog # EM-3150
Supplier:  MilliporeSigma
Supplier Number:  3150-950ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    950 ml
  • Cat. No.:
    EM-3150
  • Supplier no.:
    3150-950ML

Specifications

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
REACH:05-2114614906-45
 :GHS06
 :GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826