Anti-Beta arrestin 2 Rabbit Polyclonal Antibody
77440-416
: Bioss
- Antibody Type:Primary
- Antigen Name:Beta arrestin 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:409
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:45
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77440-416
- Packaging:Vial
Members of arrestin/beta arrestin protein family are thought to participate in agonist mediated desensitization of G protein coupled receptors and cause specific dampening of cellular responses to stimuli such as hormones, neurotransmitters, or sensory signals. Arrestin beta 2, like arrestin beta 1, was shown to inhibit beta adrenergic receptor function in vitro. It is expressed at high levels in the central nervous system and may play a role in the regulation of synaptic receptors.
Type: Primary
Antigen: Beta arrestin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat