CE4 System Sterile Latex Hand-Specific Gloves, Microflex®, Ansell
These hand specific gloves are packaged with a double vacuum seal for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) controlled environments.
- Silicone-free
- Micro-textured surface enhances wet or dry grip
- Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort
- Powder-free
These gloves are washed in microprocessor-controlled, two-in-one, washer/dryer laundering systems to provide very low surface particulates, ionic extractables, and non-volatile residues.
: Lot-specific Certificate of Analysis available upon request.
: Pair-packed in individual wallets, 10 wallets per vacuum-sealed poly bag, 20 bags per case.
: These gloves contain natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
- Catalog No:
- 32916-746
- 32916-740
- 32916-752
- 32916-744
- 32916-750
- 32916-748
- 32916-742
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Cuff Thickness:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- Finger Thickness:
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- Palm Thickness:
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil