Anti-CYP 3A4, 3A7, 3A5, and 3A9 Rabbit Polyclonal Antibody
Catalog # 77438-238
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:CYP 3A4, 3A7, 3A5, and 3A9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:1576
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:55
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77438-238
- Packaging:Vial
Specifications
About this item
Cytochromes P450 are a group of heme thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH dependent electron transport pathway. It performs a variety of oxidation reactions (e.g. caffeine 8 oxidation, omeprazole sulphoxidation, midazolam 1 hydroxylation and midazolam 4 hydroxylation) of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. The enzyme also hydroxylates etoposide.
Type: Primary
Antigen: CYP 3A4, 3A7, 3A5, and 3A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat