Anti-LC3A/B+LC3B+LC3C Rabbit Polyclonal Antibody
77440-484
: Bioss
- Antibody Type:Primary
- Antigen Name:LC3A/B+LC3B+LC3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Rat,Mouse
- Size:100 µl
- Gene ID:4130
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:17/14
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77440-484
- Packaging:Vial
Probably involved in formation of autophagosomal vacuoles (autophagosomes).
Type: Primary
Antigen: LC3A/B + LC3B + LC3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse, Rat