Anti-p66 alpha/GATAD2A Rabbit Polyclonal Antibody
Catalog # 77439-348
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:p66 alpha/GATAD2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:54815
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:68
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77439-348
- Packaging:Vial
Specifications
About this item
GATAD2A (GATA Zinc Finger Domain Containing 2A) is a Protein Coding gene. Among its related pathways are Activated PKN1 stimulates transcription of AR (androgen receptor) regulated genes KLK2 and KLK3 and Development NOTCH1-mediated pathway for NF-KB activity modulation. GO annotations related to this gene include transcription factor activity, sequence-specific DNA binding and protein binding, bridging. An important paralog of this gene is GATAD2B.
Type: Primary
Antigen: p66 alpha/GATAD2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse