Anti-Integrin alpha X /CD11c Rabbit Polyclonal Antibody
Catalog # 77439-212
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin alpha X /CD11c
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Mouse
- Size:100 µl
- Gene ID:3687
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:126 kDa
- Storage Temperature:Store at −20 °C for 12 months
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. No.:77439-212
- Packaging:Vial
Specifications
About this item
This gene encodes the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles. [provided by RefSeq].
Type: Primary
Antigen: Integrin alpha X /CD11c
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse