Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TIP49A Rabbit Polyclonal Antibody
Anti-TIP49A Rabbit Polyclonal Antibody
Catalog # 77438-844
Supplier:  Bioss
Anti-TIP49A Rabbit Polyclonal Antibody
Catalog # 77438-844
Supplier:  Bioss
Supplier Number:  BS-19936R

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    TIP49A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Gene ID:
    8607
  • Storage Buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular Weight:
    50
  • Storage Temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by protein A
  • Cat. No.:
    77438-844
  • Packaging:
    Vial

Specifications

About this item

Possesses single-stranded DNA-stimulated ATPase and ATP-dependent DNA helicase (3' to 5') activity. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. RUVBL1 plays an essential role in oncogenic transformation by MYC and also modulates transcriptional activation by the LEF1/TCF1-CTNNB1 complex. May be able to bind plasminogen at cell surface and enhance plasminogen activation.
Essential for cell proliferation.

Type: Primary
Antigen: TIP49A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse