Specifications
- Antibody Type:Primary
- Antigen Name:Integrin alpha E2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:3682
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:110/128
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Cat. No.:77439-544
- Packaging:Vial
Specifications
About this item
CD103/Integrin alpha E is a new member of the integrin series of adhesion molecules containing the beta7 subunit. This antigen defines a developmentally important subset of T cells, namely mucosal T cells including all IEL (intraepithelial lymphocytes) and ~20% of lamina propria T cells. Expression of CD103 is more restricted outside these mucosal organs, appearing at lower levels on T cell subsets of the lymph node, dendritic epidermis and periphery. In non-epithelial CD103+ T cells there is a bias toward expression on CD8+ cells. Integrin alpha E is a heterodimer comprising a 150kDa alpha chain and a 120kDa beta chain. Integrin alpha E is expressed by approximately 1% of peripheral blood lymphocytes, activated T lymphocytes and by hairy cell leukaemia cells. The antigen is also expressed by intraepithelial lymphocytes.
Type: Primary
Antigen: Integrin alpha E2
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat