Anti-Splicing factor 1 (Ser82) Rabbit Polyclonal Antibody
Catalog # 77438-304
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Splicing factor 1 (Ser82)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG (H, L)
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µl
- Gene ID:7536
- Storage Buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular Weight:0
- Storage Temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Cat. No.:77438-304
- Packaging:Vial
Specifications
About this item
This gene encodes a nuclear pre-mRNA splicing factor. The encoded protein specifically recognizes the intron branch point sequence and is required for the early stages of spliceosome assembly. Alternate splicing results in multiple transcript variants. [provided by RefSeq, May 2010]
Type: Primary
Antigen: Splicing factor 1 (Ser82)
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse