Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-METTL3 Rabbit Polyclonal Antibody
  77438-280
 :  Bioss
Anti-METTL3 Rabbit Polyclonal Antibody
  77438-280
 :  Bioss
 :  BS-17609R

 

  • Antibody Type:
    Primary
  • Antigen Name:
    METTL3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Rat
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Gene ID:
    56339
  • Storage Buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular Weight:
    64
  • Storage Temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by Protein A
  • Cat. No.:
    77438-280
  • Packaging:
    Vial

 

 

N6-methyltransferase that methylates adenosine residues of some mRNAs. N6-methyladenosine (m6A), which is present at internal sites of some mRNAs, may play a role in the efficiency of mRNA splicing, transport or translation.

Type: Primary
Antigen: METTL3
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Rat