


Zinc ≥99.8% (by EDTA titration), granular, BAKER ANALYZED® ACS (30 Mesh), J.T.Baker®
:7440-66-6
:
:Zn
:MFCD00011291
:65.39 g/mol
:12,10255
:908 °C (1013 hPa)
:420 °C
:7.14 g/cm³ (20 °C)
:Management of Change (MOC) category = N
:231-175-3
:GHS07
:GHS08
:GHS09
:Ambient
- Assay (Zn) (by EDTA titrn):≥99.8 %
- Iron (Fe):≤0.01 %
- Lead (Pb):≤0.01 %
- Suitability for Arsenic Determination:Passes Test
- Trace Impurities - Arsenic (As):≤0.100 ppm
- Mesh - on U.S. No. 30 Sieve:≤5 %
- Mesh - Thru U.S. No. 50 Sieve:≤30 %