


Acetic acid glacial 99.5-100.5%, AR Select® ACS for trace analysis, Macron Fine Chemicals™
Catalog # MK881714
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
Specifications
- Pack type:Glass bottle, clear, Mall
- Size:500 ml
- Cat. No.:MK881714
- Supplier no.:8817-14
Specifications
Specification Test Results
- ACS - Assay (CH₃COOH):99.5 - 100.5 %
- ACS - Assay (by GC, corrected for water):≥ 99.7 %
- ACS - Acetic Anhydride ((CH₃CO)₂O):≤ 0.01 %
- ACS - Arsenic and Antimony (as As):≤ 0.005 ppm
- ACS - Chloride (Cl):≤ 0.0001 %
- ACS - Color (APHA):≤ 10
- Dilution Test:Passes Test
- ACS - Heavy Metals (by ICP-OES):≤ 5 ppm
- ACS - Residue after Evaporation:≤ 0.001 %
- ACS - Substances Reducing Dichromate:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Sulfate (SO₄):≤ 0.0001 %
- ACS - Titrable Base (meq/g):≤ 0.0004
- Determine by Flame Photometry & ICP - Aluminum (Al):≤ 100 ppb
- Determine by Flame Photometry & ICP - Barium (Ba):≤ 100 ppb
- Determine by Flame Photometry & ICP - Beryllium (Be):≤ 10 ppb
- Determine by Flame Photometry & ICP - Bismuth (Bi):≤ 10 ppb
- Determine by Flame Photometry & ICP - Boron (B):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Cadmium (Cd):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Calcium (Ca):≤ 500.0 ppb
- Determine by Flame Photometry & ICP - Chromium (Cr):≤ 100 ppb
- Determine by Flame Photometry & ICP - Cobalt (Co):≤ 50 ppb
- Determine by Flame Photometry & ICP - Copper (Cu):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Gallium (Ga):≤ 500 ppb
- Determine by Flame Photometry & ICP - Germanium (Ge):≤ 500 ppb
- Determine by Flame Photometry & ICP - Gold (Au):≤ 500 ppb
- Determine by Flame Photometry & ICP - Iron (Fe):≤ 100 ppb
- Determine by Flame Photometry & ICP - Lead (Pb):≤ 500.0 ppb
- Determine by Flame Photometry & ICP - Lithium (Li):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Magnesium (Mg):≤ 100 ppb
- Determine by Flame Photometry & ICP - Manganese (Mn):≤ 50 ppb
- Determine by Flame Photometry & ICP - Mercury (Hg):≤ 500.0 ppb
- Determine by Flame Photometry & ICP - Molybdenum (Mo):≤ 10 ppb
- Determine by Flame Photometry & ICP - Nickel (Ni):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Potassium (K):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Silicon (Si):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Silver (Ag):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Sodium (Na):≤ 500.0 ppb
- Determine by Flame Photometry & ICP - Strontium (Sr):≤ 100 ppb
- Determine by Flame Photometry & ICP - Thallium (Tl):≤ 50 ppb
- Determine by Flame Photometry & ICP - Tin (Sn):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Vanadium (V):≤ 10 ppb
- Determine by Flame Photometry & ICP - Zinc (Zn):≤ 100 ppb
- Determine by Flame Photometry & ICP - Zirconium (Zr):≤ 10 ppb
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:Management of Change (MOC) category = N
REACH:01-2119475328-30
:GHS02
:GHS05
:GHS07
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152