Order Entry
Puerto Rico
ContactUsLinkComponent
Dihydrogen hexafluorozirconate 45% (w/v) in aqueous solution
Dihydrogen hexafluorozirconate 45% (w/v) in aqueous solution
Catalog # 77415-726
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Dihydrogen hexafluorozirconate 45% (w/v) in aqueous solution
Catalog # 77415-726
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC4812-1G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    1 g
  • Cat. No.:
    77415-726

Specifications

About this item

 :
 :205.215 Da
Storage Temperature:Ambient
CAS Number:12021-95-3
Formula:H₂ZrF₆
MDL Number:MFCD00082965