Order Entry
Puerto Rico
ContactUsLinkComponent
Acetonitrile, low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®
Acetonitrile, low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®
Catalog # JT9018-3
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Acetonitrile, low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®
Catalog # JT9018-3
Supplier Number:  9018-03
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    4 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    Chem21 solvent selection guide recommended by default
  • Cat. No.:
    JT9018-3
  • Supplier no.:
    9018-03

Specifications

Specification Test Results

  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:
    ≤0.010
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤190
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.3 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Appearance:
    Passes Test
  • Assay (CH₃CN) (by GC, corrected for water):
    ≥99.8 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤1.0 ppm
  • Titrable Acid (µeq/g):
    ≤0.8
  • Titrable Base (µeq/g):
    ≤0.6
  • Water (by KF, coulometric):
    ≤10 ppm

Specification Test Results

About this item

 :
 :41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Melting Pt:-45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
 :Management of Change (MOC) category = N
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878