ContactUsLinkComponent
Glow Writer™ Autoradiography Marking Pen, Diversified Biotech
Catalog # 89024-859
Small Business Enterprise
undefined
Glow Writer™ Autoradiography Marking Pen, Diversified Biotech
Catalog # 89024-859
Supplier Number:  GLWR-1000
Small Business Enterprise
Specifications
  • Description:
    Glow Writer Pen
  • Cat. No.:
    89024-859
  • Supplier no.:
    GLWR-1000
Specifications
About this item
Phosphorescent fine-point marking pen is ideal for annotating and marking chemiluminescent and isotope-labeled substrates for X-ray film autoradiography.
Delivers an optimum concentration of marking ink that is excited by room light and easy-to-read upon exposure to X-ray film. Contains a short afterglow life of approximately 2 to 3 minutes to guarantee clear visibility upon exposure.