Ammonium fluoride 40% in aqueous solution, CMOS for microelectronic, J.T.Baker®
Catalog # JT0702-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: RegulatedCertificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle, pressure bottom
- Size:7 lbs.
- Cat. No.:JT0702-5
- Supplier no.:0702-05
Specifications
Specification Test Results
- Assay (NH₄F):39.0 - 41.0 %
- Color (APHA):≤10
- pH of 1% Solution at 25°C:6.0 - 7.5
- Residue after Ignition:≤5 ppm
- Chloride (Cl):≤2 ppm
- Insoluble Matter:≤0.5 ppm
- Nitrate (NO₃):≤5 ppm
- Oxidizable Species (as SO₃):≤1 ppm
- Phosphate (PO₄):≤0.20 ppm
- Sulfate (SO₄):≤1 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Trace Impurities - Antimony (Sb):≤10.0 ppb
- Trace Impurities - Arsenic (As):≤20.0 ppb
- Arsenic and Antimony (as As):≤30.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤5.0 ppb
- Trace Impurities - Bismuth (Bi):≤50 ppb
- Trace Impurities - Boron (B):≤20.0 ppb
- Trace Impurities - Cadmium (Cd):≤20.0 ppb
- Trace Impurities - Calcium (Ca):≤10.0 ppb
- Trace Impurities - Chromium (Cr):≤10.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤50 ppb
- Trace Impurities - Germanium (Ge):≤40.0 ppb
- Trace Impurities - Gold (Au):≤20 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Lead (Pb):≤50 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤20.0 ppb
- Trace Impurities - Molybdenum (Mo):≤50.0 ppb
- Trace Impurities - Nickel (Ni):≤50 ppb
- Trace Impurities - Niobium (Nb):≤20.0 ppb
- Trace Impurities - Potassium (K):≤100 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤50.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤50.0 ppb
- Trace Impurities - Thallium (Tl):≤20.0 ppb
- Trace Impurities - Tin (Sn):≤50 ppb
- Trace Impurities - Titanium (Ti):≤50.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤20.0 ppb
- Trace Impurities - Zirconium (Zr):≤5.0 ppb
- Particle Count - 0.5 µm and greater:≤100 par/ml
- Particle Count - 1.0 µm and greater:≤25 par/ml
Specification Test Results
About this item
:
:37.04 g/mol
ADR:6.1,III
:Management of Change (MOC) category = R
EINECS:235-185-9
UN:3287
:GHS06
Storage Temperature:Ambient
CAS Number:12125-01-8
Formula:NH₄F
MDL Number:MFCD00011423