N-Acetyl-DL-tryptophan ∼99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:87-32-1
:
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00005644
:246.27 g/mol
Melting Pt:205…206 °C
Storage Temperature:Refrigerator
EINECS:201-739-3
Specification Test Results
- Identity Test:Passes
- Purity:~99%
- FTIR:Conforms to Standard
- Melting Point:200 - 210°C
- Loss on Drying:≤0.5%
- UV/VIS: λmax (water):280 ± 2 n
- Optical Rotation (c=1, 1N NaOH, 24 hrs.):0° ± 2