Order Entry
Puerto Rico
ContactUsLinkComponent
 
L(+)-Asparagine monohydrate 99.0-101.0% (dry basis), EMPROVE® EXPERT Ph. Eur. suitable for use as excipient, SAFC®
Catalog # EM1.01565.5000
Supplier:  MilliporeSigma
CAS Number:  
L(+)-Asparagine monohydrate 99.0-101.0% (dry basis), EMPROVE® EXPERT Ph. Eur. suitable for use as excipient, SAFC®
Catalog # EM1.01565.5000
Supplier:  MilliporeSigma
Supplier Number:  1.01565.5000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly drum
  • Size:
    5 kg
  • Cat. No.:
    EM1.01565.5000
  • Supplier no.:
    1.01565.5000

Specifications

Specification Test Results

  • Assay (perchloric acid titration, calculated on dried substance):
    99.0 - 101.0 %
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    white to almost white crystalline powder or colorless crystals
  • Appearance of solution (20 g/l, CO₂-free water):
    clear and colorless
  • Spec. rotation (α 20/D, 100 g/l, hydrochloric acid 3 mol/l, calc. on dried substance):
    +33.7 to +36.0
  • pH (20 g/l, CO₂-free water):
    4.0 - 6.0
  • Chloride (Cl):
    ≤ 200 ppm
  • Sulphate (SO₄):
    ≤ 200 ppm
  • Fe (Iron):
    ≤ 10 ppm
  • NH₄ (Ammonium):
    ≤ 0.1 %
  • Aspartic acid:
    ≤ 0.5 %
  • Ninhydrin - positive substances (TLC) (Glutamic acid):
    ≤ 0.5 %
  • Ninhydrin - positive substances (TLC) (any other impurity):
    ≤ 0.5 %
  • Ethanol (HS-GC):
    ≤ 5000 ppm
  • Other residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Loss on drying (130 °C, 3 h):
    10.5 - 12.5 %
  • Bacterial endotoxins:
    ≤ 3.0 I.U./g

Specification Test Results

About this item

 :
 :150.13 g/mol
Merck Index:14,00837
Melting Pt:215…217 °C
Density:1.567 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-735-9
CAS Number:5794-13-8
Formula:C₄H₈N₂O₃·1H₂O
MDL Number:MFCD00151038