Order Entry
Puerto Rico
ContactUsLinkComponent
 
Pop-Up Tube Rack, 50 ml
  77395-428
undefined
Pop-Up Tube Rack, 50 ml
  77395-428
 :  HS24319

 

  • Color:
    Green
  • Material:
    PP
  • Cat. No.:
    77395-428
  • For tubes:
    50 ml tubes, 18 wells (30 mm well Ø), 3×6 Array

 

 

Collapse for compact storage and 'pop-up' for quick use.

  • Designed to save space
  • Easy to open, holds 15 and 50 ml tubes
  • Easy to close, stackable for compact storage
  • Alphanumeric well identification for quick location and retrieval of samples

The Pop-Up rack collapses for compact storage and pops up for quick use.