Tube Rack, One-Well Wire, for 50 ml Tubes
77395-368
- Color:Yellow
- Material:Carbon steel
- Cat. No.:77395-368
- For tubes:50 ml tubes, 1 Well (29 mm well Ø)
One-well wire tube rack for 50 ml tubes.
- HDPE-coated steel wire
- Holds conical and round bottom tubes
- Open design enables easy identification of samples
- Base ring keeps tubes in place
One-well wire tube rack holds tubes for small space requirements. The rack accommodates conical and round-bottom tubes.