ContactUsLinkComponent
Acrylic Three-In-One Dispenser, Counter or Wall Mount
Acrylic Three-In-One Dispenser, Counter or Wall Mount
Catalog # 77395-340
Supplier:  VWR International
undefined
Acrylic Three-In-One Dispenser, Counter or Wall Mount
Catalog # 77395-340
Supplier:  VWR International
Specifications
  • Color:
    Gray transparent
  • Description:
    Three-in-one dispenser
  • Cat. No.:
    77395-340
Specifications
About this item
Three separate lift-up lids make loading and dispensing supplies easy.
  • Three-compartment dispenser can be used to organize an assortment of PPE supplies
  • Middle compartment has reach-in hole for dispensing
  • Durable acrylic is see-through so product levels are always visible
  • Unit includes screws for wall mounting or can be set on a counter
This three-compartment dispenser can be used to organize safety glasses, earplugs, soft covers (including bouffant caps, beard covers, shoe covers), or any small items that need to be kept within reach. The unit includes screws for wall mounting or can be set on a counter.