Flex Hold Universal Glove Box Holder
Catalog # 77395-238
Supplier: Heathrow Scientific
Specifications
- Color:Clear
- Material:PC/PETG
- Cat. No.:77395-238
- No. of compartments:4
Specifications
About this item
Glove box holder ideal for gloves, face masks, shoe covers, and more.
- Adjustable universal fit
- Easy to clean, with a low profile
- Polycarbonate slides/shelves, PETG back/bottom
- Wall or door-mountable (mounting hardware included)
- Adaptable, store different size boxes all in one holder
The flex hold universal glove box holder is ideal for holding different boxed sizes of gloves, mop caps, ear plugs, face masks, or shoe covers. It features adjustable holding points to provide true flexibility, holding up to four different boxed protective products and other lab essentials (size dependent).