Order Entry
Puerto Rico
ContactUsLinkComponent
Nalgene® CryoBox™, SYSTEM 100™, Thermo Scientific
Nalgene® CryoBox™, SYSTEM 100™, Thermo Scientific
Catalog # 66008-714
   Low Manufacturing Impact
undefined
Nalgene® CryoBox™, SYSTEM 100™, Thermo Scientific
Catalog # 66008-714
Supplier Number:  5026-1010
   Low Manufacturing Impact

Specifications

  • Dimensions:
    133L×133W×52H mm (51/4×51/4×21/16")
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Cat. No.:
    66008-714
  • Supplier no.:
    5026-1010

Specifications

About this item

These CryoBox™ containers are designed for ultralow temperature storage of one hundred SYSTEM 100™ cryogenic vials (see 66008-708 series), other vials of similar size, externally-threaded vials, and most internally-threaded vials

They increase freezer storage capacity by 23% over boxes that hold 81 vials. Constructed of polycarbonate. 10 x 10 array. A transparent lid has printed grid for inventory purposes. It permits viewing of contents and is keyed to prevent misalignment.

Temperature Range: –196 to 121 °C (–321 to 250 °F)