Order Entry
Puerto Rico
ContactUsLinkComponent
Chloromonas augustae Recombinant Channelopsin 1 (from P. pastoris)
Chloromonas augustae Recombinant Channelopsin 1 (from P. pastoris)
Catalog # 77392-874
Supplier:  Cube Biotech
CAS Number:  
undefined
Chloromonas augustae Recombinant Channelopsin 1 (from P. pastoris)
Catalog # 77392-874
Supplier:  Cube Biotech
Supplier Number:  28941
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    P. pastoris
  • Species:
    Chloromonas augustae
  • Size:
    100 µg
  • Amino Acid Number:
    362
  • Protein Synonyms:
    CaChR1
  • Protein/Peptide Name:
    Channelopsin 1
  • Purity:
    >98%
  • Molecular Weight:
    40661 Da
  • Sequence:
    MDTLAWVARE LLSTAHDATP ATATPSTDHS TPSTDHGSGE TFNVTITIGG GHHGGHAGPV DNSIVIGGID GWIAIPAGDC YCAGWYVSHG SSFEATFAHV CQWSIFAVCI LSLLWYAWQY WKATCGWEEV YVCCIELVFI CFELYHEFDS PCSLYLSTAN IVNWLRYSEW LLCCPVILIH LSNVTGLSDD YGRRTMGLLV SDIATIVFGI TAAMLVSWPK IIFYLLGFTM CCYTFYLAAK VLIESFHQVP KGICRHLVKA MAITYYVGWS FFPLIFLFGQ SGFKKISPYA DVIASSFGDL ISKNMFGLLG HFLRVKIHEH ILKHGDIRKT THLRIAGEEK EVETFVEEED EDHHHHHHHH HH
  • Cat. No.:
    77392-874

Specifications

About this item

Channelrhodopsin 1 is a 7-transmembrane protein that acts as a light-driven cation channel in eukaryotic green algae.

  • Alternative names: Channelrhodopsin 1 (CaChR1)
  • UniProt Number: G8HKA1
  • Protein class: 7-transmembrane protein
  • Organism: Chloromonas augustae (Chlamydomonas augustae)
  • Sequence, explained: Wild-type sequence, N-terminal membrane-spanning domain (362 of 715 aa), 10× His-tag
  • Affinity tags: His-tag (C-terminus)
  • Expression host: Pichia pastoris (yeast)
  • Buffer composition: 100 mM NaCl, 20 mM HEPES pH 7.4, 0.03% dodecyl maltoside (DDM)
  • Purified via: PureCube Ni-NTA Agarose
  • Absorbance: Extinction coefficient at 518 nm: 36000 M⁻¹cm⁻¹; Extinction coefficient at 590 nm: 54000 M⁻¹cm⁻¹
  • Activity: Binding of ligand all-trans retinal, covalently bound to a lysine residue
  • Evaluation of UV-VIS spectrum: absorbance ratio 280/518 nm = 2.2 (additional vibronic band at 480 nm)
  • Function: Photoreceptor protein undergoing a photocycle, Retinal protein, light-driven cation channel

Due to Channelrhodopsin 1's covalently bound colored ligand, it is ideal for use as a control membrane protein, for example, for crystallization assays and biochemical/biophysical experiments. We provide channelrhodopsin 1 of high purity, purified from yeast as a His-tagged protein.