Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MSP1D1 dH5 (from E. coli)
Human Recombinant MSP1D1 dH5 (from E. coli)
Supplier:  CUBE BIOTECH
Certificates
undefined
Human Recombinant MSP1D1 dH5 (from E. coli)
Supplier:  CUBE BIOTECH

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Storage of lyophilized copolymer: −20 °C for several years; Storage of reconstituted protein: 2...8 °C for several day
  • Amino Acid Number:
    168
  • Protein Synonyms:
    Membrane Scaffold Protein 1D1 dH5
  • Protein/Peptide Name:
    MSP1D1 dH5
  • Purity:
    >90% (Determined by SDS-PAGE)
  • Molecular Weight:
    19488 Da
  • Sequence:
    GSTFSKLREQ LGPVTQEFWD NLEKETEGLR QEMSKDLEEV AKVQPYLDDF QKKWQEEMEL RQKVEPLGEE MRDRARAHVD ALRTHLAPYS DELRQRLAAR LEALKENGGA RLAEYHAKAT EHLSTLSEKA KPALEDLRQG LLPVLESFKV SFLSALEEYT KKLNTQ
  • Shipping Temperature:
    RT

Specifications

Related Information