PureCube Fe-NTA MagBeads
Supplier: Cube Biotech
Certificates
About this item
PureCube Fe-NTA MagBeads were developed to enrich phosphorylated proteins. They can also be used to purify His-tagged proteins as a secondary function.
- Usage:
- -Enrichment of phosphopeptides
- -Specific binding and purification of 6× His-tagged proteins
- Specificity:
- -Affinity to His-tagged proteins
- -Affinity to phosphorylated biomolecules
- Bead ligand: Fe-NTA
- Bead size: 30 µm
- Filling quantity: Delivered as a 25% suspension
- Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
- pH stability: 2 to 14
- Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
The affinity matrix is based on 6% cross-linked agarose that was magnetized. The bead ligand is Fe-NTA and the particle diameter is 30 µm. They are very homogeneous in size, yielding a high degree of reproducibility. The phosphopeptide enrichment can be automated.
As an alternative, we also offer PureCube Fe-NTA Agarose resin and said resin pre-packed in cartridges/columns.
Frequently Bought Together