Specifications
- Cat. No.:77392-702
- Contents:MSP1E3D1-His lyophilized protein 2 mg
Dimyristoyl-glycerol-phosphocholine (DMPC) 5.09 mg
Sodium cholate 20 mg
Specifications
About this item
This product is a MSP nanodisc assembly kit. It contains a vial of lyophilized membrane scaffold protein MSP1E3D1 (His-tagged) and a vial of DMPC or POPC phopshospholipid to assemble a MSP nanodisc.
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: 277
- Molecular mass: 32600 Da
- Extinction coefficient (in water) ε280: 29400 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Sequence (Human origin): GHHHHHHH DYDIPTT ENLYFQG STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1E3D1 (His-tagged) creates nanodiscs of ∼12 to 14 nm in diameter.
With this assembly kit both cellular as well as cell-free expressed proteins can be stabilized in MSP nanodiscs. Just use the corresponding protocol for each purpose (see below). The MSP protein contains a His-tag, keep that in mind when choosing the affinity tag for your membrane protein.