Order Entry
Puerto Rico
ContactUsLinkComponent
Nanodisc MSP1D1 POPC
Nanodisc MSP1D1 POPC
Catalog # 77392-724
Supplier:  CUBE BIOTECH
CAS Number:  
undefined
Nanodisc MSP1D1 POPC
Catalog # 77392-724
Supplier:  CUBE BIOTECH
Supplier Number:  26333
CAS Number:  

Specifications

  • Size:
    50 µl
  • Cat. No.:
    77392-724

Specifications

About this item

This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.

  • Origin of protein: Human
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 201
  • Molecular mass: 23329 Da
  • Extinction coefficient (in water) ε280: 25440 M⁻¹ cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (Human origin): GLKLLDNWDSVTSTFSKLREQLG PVTQEFWDNLE KETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFK VSFLSALEEY TKKLNTQ
  • Affinity tag: None

Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 creates nanodiscs of ∼9 to 10 nm in diameter.

Delivery: Shipping temperature: Dry-Ice; storage of reconstituted protein: −80 °C, avoid freeze-thaw cycles.