Order Entry
Puerto Rico
ContactUsLinkComponent
Nanodisc Assembly Kit MSP1D1 dH5-His POPC
Nanodisc Assembly Kit MSP1D1 dH5-His POPC
Catalog # 77392-692
Supplier:  CUBE BIOTECH
CAS Number:  
undefined
Nanodisc Assembly Kit MSP1D1 dH5-His POPC
Catalog # 77392-692
Supplier:  CUBE BIOTECH
Supplier Number:  26223
CAS Number:  

Specifications

  • Cat. No.:
    77392-692
  • Contents:
    MSP1D1ΔH5-His lyophilized protein 2 mg; Palmitoyl-oleoyl-phosphatidylcholine (POPC) 3.12 mg; Sodium cholate 2×20 mg

Specifications

About this item

This product is a MSP nanodisc assembly kit. It contains a vial of lyophilized membrane scaffold protein MSP1D1 dh5 (His-tagged) and a vial of DMPC or POPC phopshospholipid to assemble a MSP nanodisc.

  • Origin of MS protein: Human
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 168
  • Molecular mass: 19488 Da
  • Extinction coefficient (in water) ε280:18450 M⁻¹ cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
  • Sequence (1-Letter Code): GSTFSKLREQ LGPVTQEFWD NLEKETEGLR QEMSKDLEEV AKVQPYLDDF QKKWQEEMEL RQKVEPLGEE MRDRARAHVD ALRTHLAPYS DELRQRLAAR LEALKENGGA RLAEYHAKAT EHLSTLSEKA KPALEDLRQG LLPVLESFKV SFLSALEEYT KKLNTQ
  • Affinity tag: None

Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 dh5 (His-tagged) creates nanodiscs of ∼7 to 8 nm in diameter.

With this assembly kit both cellular as well as cell-free expressed proteins can be stabilized in MSP nanodiscs. Just use the corresponding protocol for each purpose (see below). The MSP protein contains a His-tag, keep that in mind when choosing the affinity tag for your membrane protein.

Delivery: Shipping temperature: Room temperature; storage of lyophilized copolymer: −20 °C for several years; storage of reconstituted protein: 2 - 8 °C for several day.