PureCube Compact Cartridge Ni-NTA
Supplier: CUBE BIOTECH
Certificates
About this item
The PureCube Ni-NTA Compact cartridge is an IMAC chromatography column prepacked with PureCube Ni-NTA resin.
- Usage: Specific binding and purification of 6× His tagged proteins
- Specificity: Affinity to His tagged proteins
- Binding capacity: >80 mg/ml
- Bead Ligand: Ni-NTA
- Bead size: 40 μm
- Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
- Filling quantity: Delivered as a 50% suspension
- Required equipment and materials:
- FPLC hardware like e.g. ÄKTA
- Binding Buffer
- Elution Buffer
Choose between two sizes:1 ml bed volume and 5 ml bed volume. Please have a look at the given features table to ensure that the Cartridge/Column is compatible with your FPLC hardware like ÄKTA. If not, we recommend buying the sole loose PureCube Ni-NTA resin and packing the columns yourself. Alternatively, we offer our PureCube Ni-NTA products as magnetic beads if you prefer this IMAC method.