About this item
Versatile Vortex-Genie 2 mixes or shakes fluids in any type container with speed and precision.
- 220 to 230 V model
This Vortex-Genie has unmatched reliability and multi-task versatility
The 2T has timer function, both Touch and Hands-Free mode automatically turns the unit off after set time. Touch Mode: 1 to 60 seconds. Hands-Free (Switch activated Start/Stop) Mode: 1 to 60 minutes, or continuous un-timed mode. Unit automatically stops at set time – in Touch Mode or Hands-Free (switch activated Start/Stop) Mode. Accuracy ±5%. Repeatability ±1%.
It features shaking or vortexing action. Variable speed control allows slow speed shaking action up to high speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing to choose the correct action for application. A broad range of attachments of various shapes, sizes, and materials allows for the mixing of almost all common tubes or container for hands-free or touch operation. A 3-position power switch lets to select continuous action or 'Touch On' control. With the true 'Hands- Free' operation of continuous action, lab operators can greatly increase lab efficiency. Tough metal housing combined with 'just enough weight', provides a stable platform for all types of mixing.