Genie® MicroPlate, 230 V
Supplier: Electron Microscopy Sciences
Certificates
About this item
High Speed Action For Increased Mixing Efficiency In Every Well!
- 230 V model
- Aggressive mixing speed from 750-3200 rpm for complete and uniform mixing in any microplate format
- May be used to shake any single microplate
- Multi capacity microplate mixer is also available (Multi-MicroPlate Genie)
- Small vortexing orbit of 1.0 mm for thorough mixing regardless of sample viscosity
- Speed and orbit combine to offer true vortexing action in each well of a microplate
- Accepts most microplate types within the recommendations of the Society for Biomolecular Screening (SBS), even in 384-well formats
- Built-in timer operates from 1 to 60 minutes or continuous for hands-free use
- May be used in cold rooms and incubators for temperature sensitive applications
Additional accessories for the Microplate Genie® available.
Caution: 230 V model. Corresponding models with voltage suitable for use in US (102093-352 or 102093-354) is also available, please see separate product page.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.