Order Entry
Puerto Rico
ContactUsLinkComponent
6-Amino-3-chloro-2-fluorobenzoic acid 96%
Toxic
6-Amino-3-chloro-2-fluorobenzoic acid 96%
Catalog # 77415-568
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Toxic
6-Amino-3-chloro-2-fluorobenzoic acid 96%
Catalog # 77415-568
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC450566-100MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    100 mg
  • Cat. No.:
    77415-568

Specifications

About this item

 :
 :189.572 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:874782-54-4
Formula:C₇H₅ClFNO₂
MDL Number:MFCD03618452