Order Entry
Puerto Rico
ContactUsLinkComponent
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres) 90%
Flammable
Toxic
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres) 90%
Catalog # 77409-596
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Flammable
Toxic
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres) 90%
Catalog # 77409-596
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR40785-10G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    10 g
  • Cat. No.:
    77409-596

Specifications

About this item

 :
Storage Temperature:Refrigerator
UN:1992
ADR:3+6.1,III
 :GHS02
 :GHS06
CAS Number:1515-76-0
Formula:C₆H₈O₂
MDL Number:MFCD00008714