Order Entry
Puerto Rico
ContactUsLinkComponent
1H,1H,5H-Octafluoropentyl acrylate 97%
Irritant
Environment
1H,1H,5H-Octafluoropentyl acrylate 97%
Catalog # 77416-706
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Irritant
Environment
1H,1H,5H-Octafluoropentyl acrylate 97%
Catalog # 77416-706
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC5352-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    25 g
  • Cat. No.:
    77416-706

Specifications

About this item

 :
 :286.12 g/mol
Boiling Pt:56 °C/8 mmHg
Density:1.481 g/cm³ (25 °C)
Flash Pt:71 °C
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
Storage Temperature:Refrigerator
CAS Number:376-84-1
Formula:C₈H₆F₈O₂
MDL Number:MFCD00039279