Order Entry
Puerto Rico
ContactUsLinkComponent
4-Cyanophenyl isothiocyanate 97%
Corrosive
Irritant
4-Cyanophenyl isothiocyanate 97%
Catalog # 77410-280
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Corrosive
Irritant
4-Cyanophenyl isothiocyanate 97%
Catalog # 77410-280
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR5303-1G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    1 g
  • Cat. No.:
    77410-280

Specifications

About this item

 :
 :160.2 g/mol
Melting Pt:119…123 °C
Storage Temperature:Refrigerator
EINECS:220-323-2
UN:2923
ADR:8+6.1,III
 :GHS05
 :GHS07
CAS Number:2719-32-6
Formula:C₈H₄N₂S
MDL Number:MFCD00041085