1,3-Dimethyl-2-imidazolidinone
: 1,3-Dimethylimidazolidin-2-one
:80-73-9
:
:C₅H₁₀N₂O
:MFCD00003188
:114.15 g/mol
:224…226 °C
:8 °C
:1.056 g/cm³ (25 °C)
:95 °C
:201-304-8
:GHS05
:GHS07
:GHS08
:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.