Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ethynylcyclopropane 98%

Ethynylcyclopropane 98%

Flammable
Corrosive
Supplier:   APOLLO SCIENTIFIC
Ethynylcyclopropane 98%
Supplier:  APOLLO SCIENTIFIC
Certificates
Ethynylcyclopropane 98%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  Cyclopropylacetylene
  • CAS Number:6746-94-7
  • Formula:C₅H₆
  • MDL Number:MFCD02181090
  • Molecular Weight:66.1
  • Boiling Pt:51 °C
  • Density:0.781 g/cm³ (25 °C)
  • Flash Pt:–17 °C
  • Storage Temperature:Freezer
  • EINECS:425-430-1
  • UN:1993
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $21.64
    $21.64
    Each(1/Itm/Bx)
       $21.64

    Supplier Number: OR11148-5G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $63.85
    $63.85
    Each(1/Itm/Bx)
       $63.85

    Supplier Number: OR11148-25G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $181.09
    $181.09
    Each(1/Itm/Bx)
       $181.09

    Supplier Number: OR11148-100G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.