Order Entry
Puerto Rico
ContactUsLinkComponent
Pentafluorobenzoyl chloride 97%
Pentafluorobenzoyl chloride 97%
Catalog # 77416-128
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Pentafluorobenzoyl chloride 97%
Catalog # 77416-128
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC50171-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    25 g
  • Cat. No.:
    77416-128

Specifications

About this item

 :
 :230.52 g/mol
Boiling Pt:158…159 °C
Density:1.601 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,III
CAS Number:2251-50-5
Formula:C₇ClF₅O
MDL Number:MFCD00000657